Learn More
Molecular Probes™ Propidium Iodide Product Code.: 11519206
Molecular Probes™ Propidium Iodide
Product Code. 11519206
100 mg, 100mg
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mg
missing translation for 'unitSize'
100mg
Product Code. 11519206
Brand: Molecular Probes™ P1304MP
Edge
Propidium Iodide
Propidium iodide (PI) is a popular red-fluorescent nuclear and chromosome counterstain. Since propidium iodide is not permeant to live cells, it is also commonly used to detect dead cells in a population.
PI binds to DNA by intercalating between the bases with little or no sequence preference. In aqueous solution, the dye has excitation/emission maxima of 493/636 nm. Once the dye is bound, its fluorescence is enhanced 20- to 30-fold, the fluorescence excitation maximum is shifted ∼30–40 nm to the red and the fluorescence emission maximum is shifted ∼15 nm to the blue, resulting in an excitation maximum at 535 nm and fluorescence emission maximum at 617 nm.
PI is widely used in fluorescence microscopy, confocal laser scanning microscopy, flow cytometry, and fluorometry.
Learn more about propidium iodide, and propidium iodide containing products
TRUSTED_SUSTAINABILITY
Specifications
Physical Form Solution
Quantity 100 mg
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot Number1899781 Certificate TypeCertificate of Analysis Date10/08/2017 Product CodeP1304MP
Lot Number3118573 Certificate TypeCertificate of Analysis Date05/01/2025 Product CodeP1304MP
Lot Number2936401 Certificate TypeCertificate of Analysis Date30/07/2024 Product CodeP1304MP
Lot Number2916420 Certificate TypeCertificate of Analysis Date28/06/2024 Product CodeP1304MP
Lot Number2797329 Certificate TypeCertificate of Analysis Date02/12/2023 Product CodeP1304MP
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Product Title
Molecular Probes™ Propidium Iodide >