Learn More
PROTAC ERα Degrader-2, MedChemExpress Product Code.: 16410847
PROTAC ERα Degrader-2, MedChemExpress
Product Code. 16410847
50 mg, 50mg
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16410847
Brand: Medchem Express HY111846
MedChemExpress PROTAC ERα Degrader-2 comprises a IAP ligand binding group, a linker and an estrogen receptor α (ERα) binding group. PROTAC ERα Degrader-2 is an ERα degrader. Maximal ERα degradation at 30 μM concentration in human mammary tumor MCF7 cells. Degradation inducers based on cIAP1 are called specific and non-genetic IAP-dependent protein erasers (SNIPERs).
PROTAC ERα Degrader-2 comprises a IAP ligand binding group, a linker and an estrogen receptor α (ERα) binding group. PROTAC ERα Degrader-2 is an ERα degrader. Maximal ERα degradation at 30 μM concentration in human mammary tumor MCF7 cells. Degradation inducers based on cIAP1 are called specific and non-genetic IAP-dependent protein erasers (SNIPERs).
Specifications
Chemical Name or Material PROTAC ERα Degrader-2
CAS 1351169-29-3
Color White
Purity Grade Notes Research
Molecular Formula C42H61N5O8
Quantity 50 mg
Solubility Information DMSO : 200 mg/mL (261.79 mM; Need ultrasonic)
SMILES C[C@@]12[C@](CC/C2=N\OCC(NCCOCCOCCNC([C@H](CC(C)C)NC([C@@H](O)[C@H](N)CC3=CC=CC=C3)=O)=O)=O)([H])[C@@]4([H])[C@](C5=CC=C(O)C=C5CC4)([H])CC1
Molecular Weight (g/mol) 763.96
Formula Weight 763.96
Percent Purity 98.02%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
PROTAC ERα Degrader-2, MedChemExpress