Learn More
Bohlender™ PTFE Boiling Stones
Brand:  Bohlender™ H972-04
 View more versions of this product
Product Code. 10524022
  • 1740.00 DKK / Each
Only null left
Estimated Shipment: 25-08-2025
Specifications
PTFE
6 mm
500 g
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Bohlender™ PTFE Boiling Stones > Diameter: 6mm