Learn More
Bohlender™ PTFE Clock Shells
Brand:  Bohlender™ A200-04
Product Code. 10247641
  • 198.00 DKK / Each
Estimated Shipment: 02-06-2025
to see stock.
Explore more special offers
Specifications
PTFE
Watch Glass
125 mm
Convex
12 mm
Product Title
Bohlender™ PTFE Clock Shells > Diameter: 125mm