Learn More
Bohlender™ PTFE Liebig Condenser
Brand:  Bohlender™ B291-02
Product Code. 10595181
  • 3445.00 DKK / Each
Only null left
Estimated Shipment: 20-08-2025
Specifications
300 mm
1 Pack
PTFE
Liebig Condenser
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Bohlender™ PTFE Liebig Condenser > Length: 300mm