Learn More
Bohlender™ BOLA™ PTFE Moon Shaped Stirrer Shafts
Use these tilting half-moon blades for stirring in round bottom flasks with ground joint necks. Bohlender™ BOLA™ PTFE Moon Shaped Stirrer Shafts offer tangential flow with little turbulence and universal chemical resistance.
1505.00 DKK - 3780.00 DKK
Specifications
For Use With (Application) | The tilting half-moon blade is ideal for stirring in round-bottom flasks with ground joint necks. |
---|---|
Quantity | 1 Each |
Type | Moon Shaped Stirrer Shaft |
Product Code | Brand | Length (Metric) | Diameter (Metric) Shaft | Standard Taper Joint | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) Shaft | Standard Taper Joint | Price | Quantity & Availability | ||||
10751421
|
Bohlender™
C376-12 |
350 mm | 10 mm | 29/32 |
1675.00 DKK
Each |
Estimated Shipment: 25-07-2025
Log in to see stock. |
||||
10761421
|
Bohlender™
C376-14 |
450 mm | 10 mm | 29/32 |
1595.00 DKK
Each |
Estimated Shipment: 28-07-2025
Log in to see stock. |
||||
10707692
|
Bohlender™
C376-16 |
510 mm | 10 mm | 29/32 |
3245.00 DKK
Each |
Estimated Shipment: 28-07-2025
Log in to see stock. |
||||
10679211
|
Bohlender™
C376-22 |
800 mm | 16 mm | 45/40 |
2775.00 DKK
Each |
Estimated Shipment: 08-08-2025
Log in to see stock. |
||||
10518462
|
Bohlender™
C376-02 |
350 mm | 8 mm | 24/29 |
2990.00 DKK
Each |
Estimated Shipment: 28-07-2025
Log in to see stock. |
||||
10696491
|
Bohlender™
C376-18 |
600 mm | 10 mm | 29/32 |
3780.00 DKK
Each |
Estimated Shipment: 18-08-2025
Log in to see stock. |
||||
10635921
|
Bohlender™
C376-08 |
450 mm | 8 mm | 29/32 |
1505.00 DKK
Each |
Estimated Shipment: 18-08-2025
Log in to see stock. |
||||
10053910
|
Bohlender™
C376-10 |
600 mm | 8 mm | 29/32 |
1690.00 DKK
Each |
Estimated Shipment: 27-08-2025
Log in to see stock. |
||||
Description
- PTFE-jacketed stainless steel shaft with tilting half-moon stirrer blade
- Double-sided groove and access for the stirrer shaft completely made of PTFE
- Temperature resistance: -200 to +250°C
- Blades are available separately and can be mounted additionally
Specifications
The tilting half-moon blade is ideal for stirring in round-bottom flasks with ground joint necks. | |
1 Each | |
Moon Shaped Stirrer Shaft |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Bohlender™ BOLA™ PTFE Moon Shaped Stirrer Shafts