Learn More
PTGS1 293T Cell Overexpression Lysate (Denatured), Abnova
Brand:  Abnova H00005742-T01.100uL
Product Code. 16189871
  • 1710.00 DKK / 100µL
Explore more special offers
This item is not returnable. View return policy
Description
Description
Western Blotting
Specifications
Specifications
293T
PTGS1 293T Cell Transient Overexpression Lysate(Denatured)
68.7
pCMV-PTGS1 full-length
100 μL
Human
Western Blot
BC029840.1
6.8
Transient overexpression cell Lysate was tested with Anti-PTGS1 antibody (H00005742-B01) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PTGS1 293T Cell Overexpression Lysate (Denatured), Abnova > 100μL