Learn More
Keystone Cleanroom Products PureClean Self-Sealing 1421B Tyvek™ Autoclave Bag with Clear Film Side and Steam Indicator, 21 to 30 in. wide
PureClean™ Self-Sealing Autoclave Bags are made from a transparent film and 1421B DuPont™ Tyvek™, which is a new grade of uncoated, untreated, flexible Tyvek™ material specifically designed for pharmaceutical use in steam sterilization applications.
20940.00 DKK - 58895.00 DKK
Specifications
Product Type | Autoclave Bag |
---|---|
Autoclavable | Autoclavable |
Certifications/Compliance | COC |
Closure Type | Safe Seal |
Color | Clear, White |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
30042300
|
Keystone Cleanroom Products
6STKRP3060SS |
30 x 60 |
23345.00 DKK
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30042284
|
Keystone Cleanroom Products
6STKRP3036SS |
30 x 36 |
48430.00 DKK
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30042275
|
Keystone Cleanroom Products
6STKRP3030SS |
30 x 30 |
46655.00 DKK
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30042312
|
Keystone Cleanroom Products
6STKRP3028SS |
30 x 28 |
43110.00 DKK
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30042322
|
Keystone Cleanroom Products
6STKRP3020SS |
30 x 20 |
34415.00 DKK
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30042181
|
Keystone Cleanroom Products
6STKRP3016SS |
30 x 16 |
34065.00 DKK
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30042236
|
Keystone Cleanroom Products
6STKRP2844SS |
28 x 44 |
51270.00 DKK
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30042252
|
Keystone Cleanroom Products
6STKRP2840SS |
28 x 40 |
46835.00 DKK
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30042269
|
Keystone Cleanroom Products
6STKRP2834SS |
28 x 34 |
41335.00 DKK
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30042204
|
Keystone Cleanroom Products
6STKRP2832SS |
28 x 32 |
40805.00 DKK
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30042190
|
Keystone Cleanroom Products
6STKRP2828SS |
28 x 28 |
40450.00 DKK
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30042187
|
Keystone Cleanroom Products
6STKRP2646SS |
26 x 46 |
38145.00 DKK
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30042195
|
Keystone Cleanroom Products
6STKRP2634SS |
26 x 34 |
37075.00 DKK
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30042265
|
Keystone Cleanroom Products
6STKRP2628SS |
26 x 28 |
35655.00 DKK
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30042223
|
Keystone Cleanroom Products
6STKRP2462SS |
24 x 62 |
58895.00 DKK
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30042182
|
Keystone Cleanroom Products
6STKRP2452SS |
24 x 52 |
57655.00 DKK
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30042268
|
Keystone Cleanroom Products
6STKRP2448SS |
24 x 48 |
46300.00 DKK
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30042330
|
Keystone Cleanroom Products
6STKRP2440SS |
24 x 40 |
42755.00 DKK
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30042260
|
Keystone Cleanroom Products
6STKRP2436SS |
24 x 36 |
38850.00 DKK
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30042298
|
Keystone Cleanroom Products
6STKRP2430SS |
24 x 30 |
34415.00 DKK
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30042308
|
Keystone Cleanroom Products
6STKRP2424SS |
24 x 24 |
42220.00 DKK
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30042297
|
Keystone Cleanroom Products
6STKRP2260SS |
22 x 60 |
51090.00 DKK
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30042237
|
Keystone Cleanroom Products
6STKRP2240SS |
22 x 40 |
40805.00 DKK
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30042233
|
Keystone Cleanroom Products
6STKRP2226SS |
22 x 26 |
20940.00 DKK
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30042309
|
Keystone Cleanroom Products
6STKRP2224SS |
22 x 24 |
26790.00 DKK
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30042230
|
Keystone Cleanroom Products
6STKRP2134SS |
21 x 34 |
34065.00 DKK
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Uncoated Tyvek™ eliminates potential of residue deposits on equipment, post sterilization
- Clear Side enables visual confirmation of dryness for sterilized parts and equipment, which is required by EU GMP Annex 1
- Convenient Self-Seal Closure eliminates the need for autoclave tape or the validation and use of a heat sealer
- Fast and consistent for equipment preparation area
- Resists tearing and punctures
- Superior microbial barrier
- Manufactured and packaged in a certified ISO 6 cleanroom
- Made in the USA
- Certificates of Conformance sent with shipment
Specifications
Autoclave Bag | |
COC | |
Clear, White | |
Steam Sterilization of Pharmaceutical Equipment | |
Non-sterile | |
Yes | |
5.3 mil |
Autoclavable | |
Safe Seal | |
Single-use | |
Tyvek | |
5.3 mil | |
121 to 125 C |