Learn More
Fisherbrand™ Prefiltration Cartridge
Treats, softens, purifies, and conditions tap water.. Fisher Scientific™ Prefiltration Cartridges are used with the Accu500 Ultrapure Water System.
841.00 DKK - 1325.00 DKK
Specifications
For Use With (Equipment) | Accu500 Ultrapure Water System |
---|---|
Height (Metric) | 254 mm |
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
15489639
|
Fisherbrand™
LABAC1012 |
Activated Carbon Cartridge |
841.00 DKK
Pack of 12 |
Estimated Shipment: 28-05-2025
Log in to see stock. |
|||||
15499639
|
Fisherbrand™
LABAT1012 |
Anti-scaling Softening Cartridge |
1325.00 DKK
Pack of 12 |
Estimated Shipment: 12-06-2025
Log in to see stock. |
|||||
Description
- Anti-Scaling Softener Cartridge treats and conditions hard water
- Activated Carbon Cartridge removes particles, organics and chlorine
Specifications
Accu500 Ultrapure Water System | |
254 mm |