Learn More
Cole-Parmer PVC Lab and Vacuum Tubing
Smooth, polished inner wall prevents buildups—ideal for general laboratory applications
Brand:  Cole-Parmer 50121-00
 View more versions of this product
Product Code. 30053886
  • 1035.00 DKK / 15m
Estimated Shipment: 25-06-2025
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
Versatile applications including peristaltic pumps, analytical instruments, incubators, gas, and drain lines. Glass-smooth inner bore design for easy clean-ups and changeovers, enhancing workflow efficiency. Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection
Specifications
Specifications
5/16 in.
7/16 in.
Translucent
PVC
50 ft.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cole-Parmer PVC Lab and Vacuum Tubing > 5/16 in., 7/16 in.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.