Learn More
Kartell™ Plastilab™ PVC Non-toxic Isoflex Laboratory Hoses
Transparent hoses with markings at every meter for ease of use.
Brand:  Kartell™
 View more versions of this product
Product Code. 10682512
  • 969.00 DKK / 20m
Estimated Shipment: 01-04-2025
to see stock.
Explore more special offers
Description
  • Non-ageing and non-oxiding
  • Resistant to both acids and alkalis
  • Autoclavable
  • Supplied in 20-meter rolls
  • Packed in PE bags
Specifications
Specifications
Autoclavable
PVC
20 m
1 Pack
6 mm
9 mm
1.5 mm