Learn More
Robin™ PVC Weighting Ring
Brand:  Robin™ YX452
Product Code. 11927115
  • 201.00 DKK / Each
Only null left
Estimated Shipment: 13-08-2025
Specifications
PVC
Weighing Ring
220 g
38 mm
1 Ea.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Robin™ PVC Weighting Ring > Diameter: 38mm; Weight: 220g