missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ Borosilicate Glass Cylindrical Conical Flask
Ideal for fermentation and culture work
Brand: Pyrex 1190/02M
This item is not returnable.
View return policy
Description
- Manufactured from Pyrex™ borosilicate glass for effective resistance to thermal shock and chemical corrosion
- White enamel marking spot
- Autoclavable

Specifications
25 mm | |
Borosilicate Glass | |
Cylindrical |
60 mL | |
70 mm | |
45 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Pyrex™ Borosilicate Glass Cylindrical Conical Flask > Capacity: 60mL
Spot an opportunity for improvement?Share a Content Correction