Learn More
PYREX™ Culture Flask Closure
For use with 1135 culture flasks, 1138/10 Fernbach flasks and 1141 baffled flasks.
Brand:  Pyrex 4540/38P
 View more versions of this product
Product Code. 12524917
  • 343.00 DKK / Pack of 10
Estimated Shipment: 28-08-2025
Explore more special offers
Description
  • Two types of closure - Anodised aluminum - Two-position polypropylene
  • Suitable for autoclaving
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polypropylene
10
Autoclavable
Documents
Product Information
Product Title
PYREX™ Culture Flask Closure >