Learn More
Pyrex™ Media-lab Bottles with Thermoplastic Polyester Cap
Manufactured from temperature and chemically resistant borosilicate glass. Pyrex™ Media-lab Bottles with Thermoplastic Polyester Cap includes chemically resistant PTFE sealing disc and pouring ring. White graduations and marking spot add easy volume assessment. Withstands hot air sterilization up to 180° C. Complies with ISO 4796.
325.91 DKK - 4095.00 DKK
Specifications
Includes | High temperature cap and pouring ring |
---|---|
Material | Borosilicate Glass |
Certifications/Compliance | ISO 4796 |
Closure Included | Yes |
Closure Material | Thermoplastic Polyester |
Product Code | Brand | Capacity (Metric) | Closure Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Closure Size | Price | Quantity & Availability | |||||
15201499
|
Pyrex
1516/01RED |
25 mL | 25 mm |
N/A
|
N/A | |||||
15211499
|
Pyrex
1516/02RED |
50 mL | 32 mm |
1310.00 DKK
Pack of 10 |
Estimated Shipment: 29-09-2025
Log in to see stock. |
|||||
15221499
|
Pyrex
1516/04RED |
100 mL | 45 mm |
1475.00 DKK
Pack of 10 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
|||||
15231499
|
Pyrex
1516/06RED |
250 mL | 45 mm |
1685.00 DKK
Pack of 10 |
Estimated Shipment: 22-09-2025
Log in to see stock. |
|||||
15241499
|
Pyrex
1516/08RED |
500 mL | 45 mm |
1895.00 DKK
Pack of 10 |
Estimated Shipment: 01-09-2025
Log in to see stock. |
|||||
15251499
|
Pyrex
1516/10RED |
1000 mL | 45 mm |
2095.00 DKK
Pack of 10 |
Estimated Shipment: 11-08-2025
Log in to see stock. |
|||||
10660191
|
Pyrex
1516/12RED |
2000 mL | 45 mm | N/A | ||||||
15901381
|
Pyrex
1516/12RED |
2000 mL | 45 mm |
4095.00 DKK
Pack of 10 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
|||||
Description
- Manufactured from temperature and chemically resistant borosilicate glass
- Bottles complete with thermoplastic polyester cap, PTFE sealing disc, and pouring ring
- Withstands hot air sterilization up to 180° C
- PTFE sealing disc resists chemical corrosion
- White graduations and marking spot add easy volume assessment
- Complies with ISO 4796
- Printed with Trace Code for downloadable batch certificate
- Batch certificate available to download from www.scilabware.com
Specifications
High temperature cap and pouring ring | |
ISO 4796 | |
Thermoplastic Polyester |
Borosilicate Glass | |
Yes | |
Cap |