Learn More
Pyrex™ Screwcap Spare Inserts
For use with Black SVL™ PBT screwcaps
Brand:  Pyrex 4513/30M
 View more versions of this product
Product Code. 12259319
  • 4925.00 DKK / Pack of 20
Estimated Shipment: 27-08-2025
Explore more special offers
Description
  • For use with PBT screw caps
  • Autoclavable up to 121°C
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Autoclavable
PTFE
30
Sealing Disc
20/Pk.
PBT screw caps
Documents
One moment while we fetch your results.
Product Title
Pyrex™ Screwcap Spare Inserts > Thread Size: 30mm