Learn More
Pyrex™ Separating Funnel with Long Delivery Stem and PTFE Key
Supplied with hollow glass stopper and interchangeable PTFE key for grease-free performance. Pyrex™ Separating Funnel with Long Delivery Stem and PTFE Key provides good control of the liquid/liquid interface.
738.00 DKK - 2205.00 DKK
Specifications
Material | Borosilicate Glass |
---|---|
Includes | Hollow Glass stopper, PTFE key |
Product Type | Separatory Funnel |
Product Code | Brand | Capacity (Metric) | Stopcock | Joint Number | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Stopcock | Joint Number | Price | Quantity & Availability | ||||
12539165
|
Pyrex
443047N |
250 mL | (Bore Size) 2.5mm | 24/29 |
954.00 DKK
Each |
Estimated Shipment: 28-11-2025
Log in to see stock. |
||||
12519165
|
Pyrex
443042N |
125 mL | (Bore Size) 1.5mm | 19/26 |
810.00 DKK
Each |
Estimated Shipment: 28-11-2025
Log in to see stock. |
||||
12559165
|
Pyrex
443054N |
500 mL | (Bore Size) 4mm | 24/29 |
1180.00 DKK
Each |
Estimated Shipment: 28-11-2025
Log in to see stock. |
||||
12599165
|
Pyrex
443065N |
2000 mL | (Bore Size) 4mm | 29/32 |
2205.00 DKK
Each |
Estimated Shipment: 27-10-2025
Log in to see stock. |
||||
12579165
|
Pyrex
443060N |
1000 mL | (Bore Size) 4mm | 29/32 |
1475.00 DKK
Each |
Estimated Shipment: 28-11-2025
Log in to see stock. |
||||
12505067
|
Pyrex
443038N |
60 mL | (Bore Size) 1.5mm | 14/23 |
738.00 DKK
Each |
Estimated Shipment: 27-10-2025
Log in to see stock. |
||||
Description
- Gilson style with long delivery stem for good control of the liquid/liquid interface
- With interchangeable PTFE key for grease-free performance
- Supplied with hollow glass stopper
Specifications
Borosilicate Glass | |
Hollow Glass stopper, PTFE key | |
Separatory Funnel |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Pyrex™ Separating Funnel with Long Delivery Stem and PTFE Key