missing translation for 'onlineSavingsMsg'
Learn More

Quickfit™ HP Rotaflo™ Straight Pattern Plain Side-arm

Side arms are 100mm in length

667.00 DKK - 3810.00 DKK

Specifications

Type Plain Side-arm Stopcock
View More Specs

Products 5
Product Code Brand Diameter (Metric) Side Arm Length (Metric) Plug Bore Price Quantity & Availability  
Product Code Brand Diameter (Metric) Side Arm Length (Metric) Plug Bore Price Quantity & Availability  
11946728
View Documents
Quickfit
HP3ST/7
10 mm 210 mm 3 mm
667.00 DKK
Each
Estimated Shipment: 08-05-2025
to see stock.
Add to basket
 
11936728
View Documents
Quickfit
HP3ST/5
Bore: 5 mm, OD: 8 mm 210 mm 3 mm
667.00 DKK
Each
Estimated Shipment: 10-06-2025
to see stock.
Add to basket
 
15517920
View Documents
Quickfit
HP6ST/10
13 mm 210 mm 6 mm
3810.00 DKK
Pack of 5
Estimated Shipment: 08-05-2025
to see stock.
Add to basket
 
12071892
View Documents
Quickfit
HP6ST/7
10 mm 210 mm 6 mm
764.00 DKK
Each
Estimated Shipment: 10-06-2025
to see stock.
Add to basket
 
12041892
View Documents
Quickfit
HP10ST/10
13 mm 225 mm 10 mm
877.00 DKK
Each
Estimated Shipment: 03-06-2025
to see stock.
Add to basket
 
Description

Description

  • Side arms are 100mm in length
Specifications

Specifications

Plain Side-arm Stopcock
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.