missing translation for 'onlineSavingsMsg'
Learn More
Lonza™ Pyrogent–5000 Kinetic Turbidimetric LAL Assay Product Code.: 11600331

Lonza™ Pyrogent–5000 Kinetic Turbidimetric LAL Assay

Product Code. 11600331
2400 Tests/Kit, Each
Click to view available options
Quantity:
2400 Tests/Kit
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11600331

Brand: Lonza™ N588

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Includes Pyrogent 5000 LAL reconstitution buffer
Quantity 2400 Tests/Kit
Total Assay Time ∼1 hr.
Content And Storage 2°C to 8°C
For Use With (Application) Gram negative bacterial endotoxin
Product Type Pyrogent–5000 Kinetic Turbidimetric LAL Assay
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lonza™ Pyrogent–5000 Kinetic Turbidimetric LAL Assay >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.