Learn More
Bellingham Stanley™ Quartz Control Plate Sets
Bellingham Stanley™ offer convenient Quartz Control Plate (QCP) sets for verifying and calibrating polarimeters. QCPs are made to the highest standard and supplied with Certificates of Calibration.
26115.00 DKK - 36990.00 DKK
Specifications
Material Quartz
For Use With (Equipment) Polarimeter
View More Specs
Products 2
Product Code Brand Type Price Quantity & Availability  
Product Code Brand Type Price Quantity & Availability  
15977945
View Documents
Bellingham Stanley™
34-39
QCP Full Set: 100°Z, 15°Z, -30°Z
36990.00 DKK
Each
Only null left
Estimated Shipment: 17-10-2025
Add to basket
16379125
View Documents
Bellingham Stanley™
34-30
QCP Set A: 100°Z, 15°Z
26115.00 DKK
Each
Description
Description
  • Convenient Quartz Control Plate (QCP) sets for verifying and calibrating polarimeters
  • Sets available with two or three optical rotation values
  • Inert with relatively low temperature co-efficient
  • Made to the highest standard and supplied with Certificate of Calibration showing traceability to PTB
  • UKAS certified
  • Optional thermal block may be used to provide mechanical contact with the instrument temperature sensor allowing the use of quartz temperature compensation
Specifications
Specifications
Quartz
Polarimeter
Documents
Product Certifications
Product Content Correction
Product Title
Bellingham Stanley™ Quartz Control Plate Sets