Learn More
Thermo Scientific Chemicals Quartz, cover slip for microscope slide, fused, 25.4x25.4x0.15-0.25mm (1.0x1.0x0.006-0.01in) Product Code.: 15483885
In Stock
Only null left
Add to basket
Thermo Scientific Chemicals Quartz, cover slip for microscope slide, fused, 25.4x25.4x0.15-0.25mm (1.0x1.0x0.006-0.01in)
Product Code. 15483885
5/Ea., 25.4 x 25.4 mm, Pack of 5
Click to view available options
Quantity:
1/Ea.
5/Ea.
Dimensions (L x W):
25.4 mm
25.4 x 25.4 mm
Unit Size:
Each
Pack of 5
Product Code. 15483885
Brand: Thermo Scientific Chemicals 043210.KJ
Only null left
In Stock
Add to basket
Only null left
Add to basket
Request Bulk or Custom Format
For use with microscope quartz slides made by fusing naturally occurring crystalline silica
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Cover slips for use with microscope quartz slides
TRUSTED_SUSTAINABILITY
Specifications
Thickness (Metric) 0.15 to 0.25 mm
Description Cover Slips for use with Microscope Quartz Slide
Quantity 5/Ea.
Dimensions (L x W) 25.4 x 25.4 mm
RUO – Research Use Only
Product Content Correction
Product Title
Thermo Scientific Chemicals Quartz, cover slip for microscope slide, fused, 25.4x25.4x0.15-0.25mm (1.0x1.0x0.006-0.01in) >