Learn More
Hellma™ Quartz Flow Through Cells Far UV
Brand:  Hellma™ 170-700-2-40
 View more versions of this product
Product Code. 11716274
  • 6625.00 DKK / Each
Estimated Shipment: 01-10-2025
Explore more special offers
Specifications
Flow-through Cell Far UV
Quartz
2 mm
Product Title
Hellma™ Quartz Flow Through Cells Far UV > Light Path: 2mm