Learn More
Hellma™ Quartz Flow Through Cells Far UV
Brand:  Hellma™ 170700-0.1-40
 View more versions of this product
Product Code. 11774296
  • 8735.00 DKK / Each
Estimated Shipment: 13-06-2025
Explore more special offers
Specifications
Flow-through Cell Far UV
Quartz
0.1 mm
Product Title
Hellma™ Quartz Flow Through Cells Far UV > Light Path: 0.1mm