Learn More
Quickfit™ 20 mm Plastic Cap Product Code.: 15748027
Estimated Shipment: 22-07-2025
to see stock.
Add to basket
Quickfit™ 20 mm Plastic Cap
Product Code. 15748027
Product Code. 15748027
Brand: Quickfit ML/CAP20
Estimated Shipment: 22-07-2025
to see stock.
Add to basket
Add to basket
Quickfit™ 20 mm Plastic Cap is a spare part for use with the ML1 microscale kit.
  • Spare screw cap for ML1 microscale kit and microscale dropping funnel
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene
Product Type Cap
For Use With (Equipment) ML1 microscale kit, thermometer adapter
Closure Size 20 mm
Diameter (Metric) 20 mm
Closure Type Screw Cap
Product Content Correction
Product Title
Quickfit™ 20 mm Plastic Cap >