Learn More
Quickfit™ Borosilicate Glass Air Condenser
Manufactured from Pyrex™ Borosilicate Glass. Quickfit™ Borosilicate Glass Air Condenser creates condensation of materials with boiling points above 150°C.
Brand:  Quickfit C2/22
Product Code. 12518865
  • 403.00 DKK / Each
Estimated Shipment: 22-09-2025
to see stock.
Explore more special offers
Description
Simple condensers for condensation of materials with boiling points above 150°C
Product Title
Quickfit™ Borosilicate Glass Air Condenser > Cone Size: 19/26; Length: 470mm; Effective Length: 400mm