Learn More
Quickfit™ Borosilicate Glass Column Packing Raschig Ring
Brand:  Quickfit FC8/06
Product Code. 12011662
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • For use with Quickfit™ plain fractionating columns
  • Raschig rings - short lengths of glass tubing
  • Supplied in 1L bags
  • Manufactured from borosilicate glass for contamination free performance
TRUSTED_SUSTAINABILITY
Specifications
Specifications
6mm
Raschig Ring
Pyrex Borosilicate Glass
Quickfit™ plain fractionating columns
Product Title
Quickfit™ Borosilicate Glass Column Packing Raschig Ring > Length: 6mm