Learn More
Quickfit™ Borosilicate Glass Iodine Flask
Complies with BS 2735
Brand: Quickfit FIB250/4
This item is not returnable.
View return policy
Description
Complies with BS 2735
- Complies with BS 2735
- For the determination of iodine
- Flask has ground socket and cup shaped top
- Stopper has extended handle which projects well above the liquid seal
Note: Stopper is not included; sold separately.
Specifications
BS 2735 | |
250 mL | |
29/32 |
Top style: Cup shaped | |
Borosilicate Glass |
Product Content Correction
Product Title
Quickfit™ Borosilicate Glass Iodine Flask > Capacity: 250mL; Socket: 29/32
Spot an opportunity for improvement?Share a Content Correction