Learn More
Quickfit™ Borosilicate Glass Medium Neck Round Bottom Flask
Complies with BS 2735
Brand:  Quickfit FR500/3M
489.25 DKK valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 11318054
  • 528.00 DKK / Pack of 2
Estimated Shipment: 13-03-2025
to see stock.
Explore more special offers
Description
Complies with BS 2735
  • With a longer neck than standard Quickfit™ flasks
  • Useful for reducing problems associated with frothing and splashing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
500 mL
Useful for reducing problems associated with frothing and splashing
Medium
Borosilicate Glass
180 mm
24/29