Learn More
Quickfit™ Borosilicate Glass Medium Neck Round Bottom Flask
Complies with BS 2735
Brand: Quickfit FR500/3U
216.60 DKK valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Complies with BS 2735
- With a longer neck than standard Quickfit™ flasks
- Useful for reducing problems associated with frothing and splashing
Specifications
500 mL | |
Useful for reducing problems associated with frothing and splashing | |
Medium |
Borosilicate Glass | |
205 mm | |
24/29 |