Learn More
Quickfit™ Borosilicate Glass Sintered Dip Tube for Dreschel Bottle
Disperse gas into bubbles with QuckFit™ sintered borosilicate glass tubes designed to fit a range of QuickFit™ Deschel bottles. Tubes come in a variety of porosity for fine control over bubble size.
Brand:  Quickfit MF48/03/250
 View more versions of this product
Product Code. 11312065
  • 682.00 DKK / Each
Only null left
Estimated Shipment: 19-09-2025
Description
Disperse gas into bubbles with QuckFit™ sintered borosilicate glass tubes designed to fit a range of QuickFit™ Deschel bottles. Tubes come in a variety of porosity for fine control over bubble size.
  • Complies with BS 2461
  • Sintered dip tube disperses gas into bubbles for greater efficiency
  • Choice of sinter porosities to achieve fine bubble size
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BS 2461
Borosilicate Glass
Bottle Heads
Stem Length: 165 mm
For model MF29/3/250 Dreschel bottle
0
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Quickfit™ Borosilicate Glass Sintered Dip Tube for Dreschel Bottle > Stem Length: 165mm; Porosity: 0