Learn More
Quickfit™ Plain Bend Delivery Receiver
Plain bend without vent
Brand:  Quickfit RA9/44
367.65 DKK valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 12069111
  • 397.00 DKK / Each
Estimated Shipment: 22-04-2025
to see stock.
Explore more special offers
Description
Quickfit™ Plain Bend Delivery Receiver adapters are available in a range of cone sizes.
  • Plain bend without vent
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Receiver Adapter
140 mm
29/32
Borosilicate Glass
5.5 in.