missing translation for 'onlineSavingsMsg'
Learn More
(R)-(-)-1-(1-Naphthyl)ethyl isocyanate, 97% Product Code.: 11328725

(R)-(-)-1-(1-Naphthyl)ethyl isocyanate, 97%

Product Code. 11328725
250 mg, 250mg
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

Product Code. 11328725

Brand: Thermo Scientific Alfa Aesar L12286.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

CAS: 42340-98-7 | C13H11NO | 197.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42340-98-7
Molecular Formula C13H11NO
Molecular Weight (g/mol) 197.24
MDL Number MFCD00004015
InChI Key GONOHGQPZFXJOJ-SNVBAGLBSA-N
Synonym r---1-1-naphthyl ethyl isocyanate, isocyanic acid r---1-1-naphthyl ethyl ester, 1-1r-1-isocyanatoethyl naphthalene, naphthalene,1-1r-1-isocyanatoethyl, 1r-1-naphthylethanisocyanate, 1-1-isocyanatoethyl naphthalene #, pubchem8100, r-nei, r-1-1-naphthyl ethyl isocyanateShow MoreShow Less
PubChem CID 170633
IUPAC Name 1-[(1R)-1-isocyanatoethyl]naphthalene
SMILES C[C@@H](N=C=O)C1=C2C=CC=CC2=CC=C1

Specifications

Density 1.118
Boiling Point 106°C to 108°C (0.16 mmHg)
Flash Point 93°C (199°F)
Refractive Index 1.6045
Quantity 250 mg
UN Number UN2206
Beilstein 4181805
Sensitivity Moisture sensitive
Optical Rotation −47° (c=3.5 in Toluene)
Formula Weight 197.24
Percent Purity 97%
Chemical Name or Material (R)-(-)-1-(1-Naphthyl)ethyl isocyanate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Product Code
Lot Number10120771 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeL12286.MD,L12286.03
Lot Number90010872 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeL12286.MD,L12286.03
Lot Number10153339 Certificate TypeCertificate of Analysis Date06/12/2021 Product CodeL12286.MD,L12286.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(R)-(-)-1-(1-Naphthyl)ethyl isocyanate, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.