missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(-)-1-Methyl-3-phenylpropylamine, 98%, Thermo Scientific Chemicals
1252.21 DKK - 4806.39 DKK
Chemical Identifiers
CAS | 937-52-0 |
---|---|
Molecular Formula | C10H15N |
Molecular Weight (g/mol) | 149.24 |
MDL Number | MFCD00145208 |
InChI Key | WECUIGDEWBNQJJ-UHFFFAOYNA-N |
Synonym | r---1-methyl-3-phenylpropylamine, r-4-phenylbutan-2-amine, 2r-4-phenylbutan-2-amine, unii-o4f6h42uwq, o4f6h42uwq, r-1-methyl-3-phenylpropylamine, r-4-phenylbutane-2-amine, 4-phenylbutan-2-amine, r, r-1-methyl-3-phenyl-propylamineShow More |
PubChem CID | 2734033 |
SMILES | CC(N)CCC1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10722212
|
Thermo Scientific Acros
414760010 |
1 g |
1252.21 DKK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11945251
|
Thermo Scientific Acros
414760050 |
5 g |
4806.39 DKK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
937-52-0 | |
149.24 | |
WECUIGDEWBNQJJ-UHFFFAOYNA-N | |
2734033 |
C10H15N | |
MFCD00145208 | |
r---1-methyl-3-phenylpropylamine, r-4-phenylbutan-2-amine, 2r-4-phenylbutan-2-amine, unii-o4f6h42uwq, o4f6h42uwq, r-1-methyl-3-phenylpropylamine, r-4-phenylbutane-2-amine, 4-phenylbutan-2-amine, r, r-1-methyl-3-phenyl-propylamineShow More | |
CC(N)CCC1=CC=CC=C1 |
Specifications
937-52-0 | |
0.9360g/mL | |
98% | |
C10H15N | |
MFCD00145208 | |
− 11.00 (20.00°C neat) | |
r---1-methyl-3-phenylpropylamine, r-4-phenylbutan-2-amine, 2r-4-phenylbutan-2-amine, unii-o4f6h42uwq, o4f6h42uwq, r-1-methyl-3-phenylpropylamine, r-4-phenylbutane-2-amine, 4-phenylbutan-2-amine, r, r-1-methyl-3-phenyl-propylamine | |
CC(N)CCC1=CC=CC=C1 | |
149.24 | |
149.24 | |
Liquid |
Colorless | |
Authentic | |
Glass bottle | |
1.5130 to 1.5150 | |
0.936 | |
− 11.00 | |
WECUIGDEWBNQJJ-UHFFFAOYNA-N | |
4-phenylbutan-2-amine | |
2734033 | |
98% | |
(R)-(-)-1-Methyl-3-phenylpropylamine |
Safety and Handling
GHS Signal Word: Danger
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- (R)-(-)-1-Methyl-3-phenylpropylamine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H311-Toxic in contact with skin.
- H412-Harmful to aquatic life with long lasting effects.
- H301-Toxic if swallowed.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
(R)-(-)-1-Methyl-3-phenylpropylamine, 98%, Thermo Scientific Chemicals