missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-3-(Dimethylamino)pyrrolidine dihydrochloride, 97%, Thermo Scientific™

Product Code. 15408925
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 864448-61-3
Molecular Formula C6H16Cl2N2
Molecular Weight (g/mol) 187.108
MDL Number MFCD08704347
InChI Key LCPKWRSLMCUOOZ-QYCVXMPOSA-N
Synonym r-+-3-dimethylamino pyrrolidine dihydrochloride, r-dimethylpyrrolidin-3-yl-amine dihydrochloride, r-n,n-dimethylpyrrolidin-3-amine dihydrochloride, 3r-n,n-dimethylpyrrolidin-3-amine dihydrochloride, r-3-dimethylaminopyrrolidine 2hcl, 3r-+-3-dimethylamino pyrrolidine 2hcl, r-3-dimethylamino pyrrolidine dihydrochloride, r-n,n-dimethyl-3-pyrrolidinamine.2hcl, r-3-dimethylaminopyrrolidine dihydrochloride, dimethyl-pyrrolidin-3-yl-amine dihydrochloride
PubChem CID 53487164
IUPAC Name (3R)-N,N-dimethylpyrrolidin-3-amine;dihydrochloride
SMILES CN(C)C1CCNC1.Cl.Cl

Specifications

Melting Point 194°C to 197°C
Quantity 1 g
Sensitivity Hygroscopic
Optical Rotation +3.1° (c=1 in Water)
Formula Weight 187.11
Percent Purity 97%
Chemical Name or Material (R)-(+)-3-(Dimethylamino)pyrrolidine dihydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'