missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human CD40 Ligand/TNFSF5 (HEK293-expressed)
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
3210.50 DKK
Specifications
Accession Number | P29965 |
---|---|
For Use With (Application) | Bioactivity |
Gene ID (Entrez) | 959 |
Name | CD40 Ligand/TNFSF5 |
Quantity | 25 μg |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18318367
|
R&D Systems™
6420-CL-025/CF |
Lyophilized from a 0.2 μm filtered solution in PBS and EDTA. |
3210.50 DKK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18310558
|
R&D Systems™
6420-CL-025 |
Lyophilized from a 0.2 μm filtered solution in PBS and EDTA with BSA as a carrier protein. |
3210.50 DKK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Measured in a cell proliferation assay using enriched human B cells in the presence of IL-4. Spriggs, M.K. et al. (1992) J. Exp. Med. 176:1543. The ED50 for this effect is 0.2-1.2 ng/mL in the presence of 20 ng/mL of Recombinant Human IL-4 (Catalog # 204-IL ) and a cross-linking antibody, Mouse Anti-Hemagglutinin/HA Peptide Monoclonal Antibody (Catalog # MAB060 ).Specifications
P29965 | |
959 | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. | |
CD40LG | |
Recombinant | |
CD40 Ligand/TNFSF5 |
Bioactivity | |
CD40 Ligand/TNFSF5 | |
Human embryonic kidney cell, HEK293-derived human CD40 Ligand/TNFSF5 protein Hemagglutinin tag YPYDVPDYA GCN4-IZ (Met1-Ile29) GGGSGGGSGGGS Human CD40 Ligand (Met113-Leu261) (N-terminus) Accession # P29965 (C-terminus) | |
<0.01 EU / 1 μg of the protein by the LAL method. | |
Unconjugated | |
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |