Learn More
R&D Systems™ Human Integrin alpha 5/CD49e (NP_002196) VersaClone cDNA
ITGA5
Brand:  R&D Systems™ RDC0447
Product Code. 18727568
  • 5645.00 DKK / 10µg
This item is not returnable. View return policy
Description
Description
The Human Integrin alpha 5/CD49e (NP_002196) VersaClone cDNA clone contains the complete ORF for Integrin alpha 5/CD49e, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Integrin alpha 5/CD49e insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
hITGA5
10 μg at 0.2 μg/μL
3163bp
Human
NP_002196
cDNA is provided in 10mM Tris-Cl, pH 8.5
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
10 μg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
R&D Systems™ Human Integrin alpha 5/CD49e (NP_002196) VersaClone cDNA > 10ug
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.
For Reasearch Use Only; Not for Use in Humans