Learn More
R&D Systems™ Human Integrin alpha L/CD11a (NP_002200) VersaClone cDNA
ITGAL
Brand: R&D Systems™ RDC0495
This item is not returnable.
View return policy
Description
The Human Integrin alpha L/CD11a (NP_002200) VersaClone cDNA clone contains the complete ORF for Integrin alpha L/CD11a, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Integrin alpha L/CD11a insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Order Info
Ships at ambient temperature.
Specifications
hITGAL/CD11a | |
10 ÎĽg at 0.2 ÎĽg/ÎĽL | |
3526bp | |
Human |
NP_002200 | |
cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 ÎĽg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
R&D Systems™ Human Integrin alpha L/CD11a (NP_002200) VersaClone cDNA > 10ug
For Reasearch Use Only; Not for Use in Humans
Spot an opportunity for improvement?Share a Content Correction