Learn More
R&D Systems™ Mouse CysTLR2 (NP_598481) VersaClone cDNA
Cysteinyl Leukotriene Receptor 2
Brand:  R&D Systems™ RDC0450
Product Code. 18481170
  • 2460.00 DKK / 10µg
Explore more special offers
Description
Description
The Mouse CysTLR2 (NP_598481) VersaClone cDNA clone contains the complete ORF for Cysteinyl Leukotriene R2/CysLTR2, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Cysteinyl Leukotriene R2/CysLTR2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
Specifications
Specifications
mCYSLTR2
10 μg at 0.2 μg/μL
943bp
Mouse
NP_598481
cDNA is provided in 10mM Tris-Cl, pH 8.5
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
10 μg
For Reasearch Use Only; Not for Use in Humans