Learn More
R&D Systems™ Mouse Relaxin R1 (NP_997617) VersaClone cDNA
Relaxin Receptor 1
Brand:  R&D Systems™ RDC0790
Product Code. 18491560
  • 5645.00 DKK / 10µg
Description
The Mouse Relaxin R1 (NP_997617) VersaClone cDNA clone contains the complete ORF for Relaxin R1/LGR7, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Relaxin R1/LGR7 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
mRXFP1
10 μg at 0.2 μg/μL
2289bp
Mouse
NP_997617
cDNA is provided in 10mM Tris-Cl, pH 8.5
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
10 μg
Certificates
Promotions
Promotions
Product Content Correction
Product Title
R&D Systems™ Mouse Relaxin R1 (NP_997617) VersaClone cDNA > 10ug
For Reasearch Use Only; Not for Use in Humans