Learn More
(R)-(-)-Gossypol acetic acid, MedChemExpress
MedChemExpress (R)-(-)-Gossypol acetic acid (AT-101 (acetic acid)) is the levorotatory isomer of a natural product Gossypol. AT-101 is determined to bind to Bcl-2, Mcl-1 and Bcl-xL proteins with Kis of 260±30 nM, 170±10 nM, and 480±40 nM, respectively.

1148.12 DKK - 3746.49 DKK
Specifications
Chemical Name or Material | (R)-(-)-Gossypol acetic acid |
---|---|
Physical Form | Solid |
CAS | 866541-93-7 |
Color | Dark Brown |
Purity Grade Notes | Research |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16452637
![]() |
Medchem Express
HY-15464A |
50 mg |
3746.49 DKK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16426548
![]() |
Medchem Express
HY-15464A |
10 mg |
1148.12 DKK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
(R)-(-)-Gossypol acetic acid (AT-101 (acetic acid)) is the levorotatory isomer of a natural product Gossypol. AT-101 is determined to bind to Bcl-2, Mcl-1 and Bcl-xL proteins with Kis of 260±30 nM, 170±10 nM, and 480±40 nM, respectively.Specifications
(R)-(-)-Gossypol acetic acid | |
866541-93-7 | |
Research | |
Cancer-programmed cell death | |
DMSO : 25 mg/mL (43.21 mM; Need ultrasonic) | |
578.61 | |
96.98% |
Solid | |
Dark Brown | |
C32H34O10 | |
AT-101 (acetic acid) (-)-Gossypol acetic acid (R)-Gossypol acetic acid | |
OC1=C2C(C=O)=C(O)C(O)=C(C(C)C)C2=CC(C)=[C@]1[C@]3=C(C)C=C4C(C(C)C)=C(O)C(O)=C(C=O)C4=C3O.CC(O)=O.[R] | |
578.61 | |
Research |
Safety and Handling
missing translation for 'shelfLife' : -20°C, protect from light, stored under nitrogen∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light, stored under nitrogen)
Recommended Storage : -20°C, protect from light, stored under nitrogen∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light, stored under nitrogen)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.