missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ RAB7L1 Recombinant Protein

2665.00 DKK - 4040.00 DKK

Specifications

Accession Number AAH02585
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 8934
Molecular Weight (g/mol) 48.07
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16133112
View Documents Promotion Details
Abnova™
H00008934-P01.10ug
10 ug
2665.00 DKK
10µg
Please to purchase this item. Need a web account? Register with us today!
16143112
View Documents Promotion Details
Abnova™
H00008934-P01.25ug
25 ug
4040.00 DKK
25µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH02585
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
48.07
8
Glutathione Sepharose 4 Fast Flow
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
RAB7L1
Human
Recombinant
Solution
Antibody Production, Protein Array, ELISA, Western Blot
8934
RAB7L1 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
MGSRDHLFKVLVVGDAAVGKTSLVQRYSQDSFSKHYKSTVGVDFALKVLQWSDYEIVRLQLWDIAGQERFTSMTRLYYRDASACVIMFDVTNATTFSNSQRWKQDLDSKLTLPNGEPVPCLLLANKCDLSPWAVSRDQIDRFSKENGFTGWTETSVKENKNINEAMRVLIEKMMRNSTEDIMSLSTQGDYINLQTKSSSWSCC
DKFZp686P1051/RAB7L
RAB7L1
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Product Certifications