missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAD9B Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-06241IR
This item is not returnable.
View return policy
Description
RAD9B Polyclonal specifically detects RAD9B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
RAD9B | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
cell cycle checkpoint control protein RAD9B, cell cycle checkpoint control protein RAD9B homolog, DNA repair exonuclease rad9 homolog B, FLJ40346, hRAD9B, MGC75426, RAD9 homolog B (S. pombe) | |
Produced in rabbits immunized with E. coli-derived Human RAD9B fragment. | |
0.1 mL | |
Cell Cycle and Replication | |
144715 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |