Learn More
STEMCELL Technologies Rapamycin
Antibiotic; mTOR pathway inhibitor; Inhibits FKBP-12
641.25 DKK - 2577.15 DKK
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15682517
![]() |
STEMCELL Technologies
73362 |
1 mg |
641.25 DKK
1mg Save Up to 383.54 DKK » |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17130212
![]() |
STEMCELL Technologies
73364 |
10 mg |
2577.15 DKK
10mg Save Up to » |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Rapamycin is a macrolide antibiotic and immunosuppressive compound that inhibits mammalian target of rapamycin (mTOR) signaling
- It acts through formation of a complex with cytosolic FK-binding protein 12 (FKBP-12), which directly binds to mTOR complex 1 (mTORC1)
- Its immunosuppressive effects are mediated through inhibition of IL-2 signaling that is critical for T-cell proliferation and activation (Gibbons et al.; Kay et al.) Rapamycin shows antifungal activity against Candida albicans and other fungi (Vézina et al.)
- Inhibits growth of MDA-MB-468 human breast cancer cells in vitro, and inhibits tumor growth in a mouse xenograft model in vivo (Akcakanat et al.)
- Induces autophagy in malignant glioma cells (Takeuchi et al.)
Specifications
53123-88-9 | |
≥95% |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
STEMCELL Technologies Rapamycin