missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAPGEF5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1081.84 DKK - 3058.99 DKK
Specifications
Antigen | RAPGEF5 |
---|---|
Concentration | 0.25 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16810324
|
Proteintech
12556-1-AP-20UL |
20 ÎĽL |
1081.84 DKK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16800324
|
Proteintech
12556-1-AP-150UL |
150 ÎĽL |
3058.99 DKK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Members of the RAS subfamily of GTPases function in signal transduction as GTP/GDP-regulated switches that cycle between inactive GDP- and active GTP-bound states. Guanine nucleotide exchange factors (GEFs), such as Repac, serve as RAS activators by promoting acquisition of GTP to maintain the active GTP-bound state and are the key link between cell surface receptors and RAS activation. Repac, also known as Rap guanine nucleotide exchange factor 5 (RAPGEF5), is a 580 amino acid protein that is expressed in brain and testis, with weaker expression in heart, placenta, lung, pancreas and small intestine. Existing as two alternatively spliced isoforms, the Repac gene is conserved in cow, mouse and rat. The gene that encodes Repac maps to human chromosome 7p15.3, with high expression of this region linking to positive response to erlotinib therapy for those with non-small-cell lung cancer.Specifications
RAPGEF5 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q8C0Q9, Q92565 | |
217944, 9771 | |
RAPGEF5 Fusion Protein Ag3229 | |
Primary | |
-20°C | |
RAPGEF5 |
0.25 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
GFR, KIAA0277, M Ras regulated Rap GEF, MR GEF, MRGEF, RAPGEF5, Related to Epac, REPAC | |
RAPGEF5 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RAPGEF5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech