missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RCOR1/CoREST Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
3439.44 DKK
Specifications
Antigen | RCOR1/CoREST |
---|---|
Dilution | Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
RCOR1/CoREST Polyclonal specifically detects RCOR1/CoREST in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
RCOR1/CoREST | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Transcription Factors and Regulators | |
PBS buffer, 2% sucrose | |
23186 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Polyclonal | |
Purified | |
RUO | |
Human | |
KIAA0071CORESTRCORREST corepressor, Protein CoREST, REST corepressor 1 | |
The immunogen is a synthetic peptide directed towards the middle region of human RCOR1/CoREST (NP_055971). Peptide sequence DEVLQEWEAEHGKEETNGPSNQKPVKSPDNSIKMPEEEDEAPVLDVRYAS | |
Affinity purified |