missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RDH11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11077-1-AP-150UL
This item is not returnable.
View return policy
Description
RHD11, a member of the short-chain dehydrogenase/reductase superfamily of oxidoreductases, is expressed at high levels in prostate epithelium, and its expression is regulated by androgens.Specifications
RDH11 | |
Polyclonal | |
Unconjugated | |
RDH11 | |
ARSDR1, CGI 82, HCBP12, MDT1, PSDR1, RALR1, RDH11, Retinal reductase 1, Retinol dehydrogenase 11, SCALD, SDR7C1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17252, 362757, 51109 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8TC12, Q9QYF1 | |
Rdh11 | |
RDH11 Fusion Protein Ag1485 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RDH11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction