Learn More
Invitrogen™ ReadyProbes™ Streptavidin/Biotin Blocking Solution (1X)
ReadyProbes Streptavidin/Biotin Blocking Solution blocks endogenous biotin, biotin receptors, and streptavidin binding sites present in many types of tissue, reducing background signal in IHC, ICC, IF, and ISH procedures.
Brand:  Invitrogen™ R37628
993.38 DKK valid until 2025-06-21
Use promo code "24112" to get your promotional price.
Product Code. 16553652
  • 1490.00 DKK / 1 set
Get 3 for 2
Use promo code 24112 at checkout
View Available Promotions
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price
Description
Description
ReadyProbes Streptavidin/Biotin Blocking Solution blocks endogenous biotin, biotin receptors, and streptavidin binding sites present in many types of tissue, reducing background signal in immunohistochemistry (IHC), immunocytochemistry (ICC), immunofluorescence (IF), and in situ hybridization (ISH) procedures. This two-component solution (a streptavidin-blocking solution and a biotin-blocking solution) is provided in a ready-to-use format—just add a few drops of the streptavidin component and incubate for 15 minutes, then wash and repeat with the biotin component. ReadyProbes Streptavidin/Biotin Blocking Solution is used after the blocking of cells/tissue with standard non-specific blocking solution (e.g., diluted serum), but before the primary antibody step.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Blocking Solution
ReadyProbes™
Approved for shipment at Room Temperature or on Wet Ice
1 kit
A kit containing 18 mL each ready-to-use (1 X) solutions of streptavidin (component A) and biotin (component B) in two clear LDPE dropper bottles.
Store (at 2°C to 8°C.
When stored as directed, product can be used for 6 months after receipt.
)
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2907382 Certificate TypeCertificate of Analysis Date09/08/2024 Catalog NumberR37628
Lot Number2881762 Certificate TypeCertificate of Analysis Date03/04/2024 Catalog NumberR37628
Lot Number2790058 Certificate TypeCertificate of Analysis Date22/11/2023 Catalog NumberR37628
Lot Number2752682 Certificate TypeCertificate of Analysis Date29/09/2023 Catalog NumberR37628
Lot Number2738267 Certificate TypeCertificate of Analysis Date02/08/2023 Catalog NumberR37628
Promotions
Promotions
  • Multiple Manufacturers
    Exclusive savings, limited-time offer.
    Promo Code: 24112
    Expires2025-06-20
    Days
    5
    Hours
    13
    Minutes
    42
    Seconds
    46
    New Offer
    Save Now
Product Content Correction
Product Title
Invitrogen™ ReadyProbes™ Streptavidin/Biotin Blocking Solution (1X) > When stored as directed, product can be used for 6 months after receipt.