Learn More
R&D Systems™ Recombinant Cynomolgus alpha-L-Iduronidase/IDUA Protein
His-tag
Brand:  R&D Systems™ 11281-GH-020
Product Code. 18768016
  • 5265.00 DKK / 20µg
Explore more special offers
This item is not returnable. View return policy
Description
Description
Measured by its ability to cleave a fluorogenic substrate, 4-Methylumbelliferyl alpha-L-iduronide. The specific activity is >15,000 pmol/min/μg, as measured under the described conditions.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
XP_005554313.1
Supplied as a 0.2 μm filtered solution in Sodium Acetate, NaCl and Glycerol.
20 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70 °C as supplied. 3 months, -20 to -70 °C under sterile conditions after opening.
Unconjugated
Enzyme Activity
M.W. Observed: 81-90 kDa, under reducing conditions, M.W. Theoretical: 72 kDa
Chinese Hamster Ovary cell line, CHO-derived cynomolgus monkey alpha-L-Iduronidase/IDUA protein Ala26-Pro653 with a C-terminal 10-His tag
<0.10 EU per 1 μg of the protein by the LAL method.
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie® Blue Staining.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
R&D Systems™ Recombinant Cynomolgus alpha-L-Iduronidase/IDUA Protein > 20 μg