missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Carbon storage regulator Protein
A DNA sequence encoding the Escherichia coli (strain K12 / DH10B) Carbon storage regulator, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7510-ec-500ug
This item is not returnable.
View return policy
Specifications
WP_000906486.1 | |
Carbon storage regulator Protein | |
Research Use Only | |
csrA | |
Recombinant Protein | |
E. coli | |
MLILTRRVGE TLMIGDEVTV TVLGVKGNQV RIGVNAPKEV SVHREEIYQR IQAEKSQQSS Y | |
Greater than 90% as determined by SDS-PAGE. |
32441215 | |
500 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |