missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Actin-related protein 2 / 3 complex subunit 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Actin-related protein 2/3 complex subunit 2, was expressed in the hosts and tags indicated.
1185.00 DKK - 15565.00 DKK
Specifications
Accession Number | NP_005722.1 |
---|---|
Gene ID (Entrez) | 10109 |
Name | Actin-related protein 2 / 3 complex subunit 2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16074641
|
enQuireBio™
QP8442-ec-10ug |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16014651
|
enQuireBio™
QP8442-ec-50ug |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16064641
|
enQuireBio™
QP8442-ec-100ug |
100 μg |
3450.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16094641
|
enQuireBio™
QP8442-ec-200ug |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16004651
|
enQuireBio™
QP8442-ec-500ug |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16084641
|
enQuireBio™
QP8442-ec-1mg |
1 mg |
15565.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_005722.1 | |
Actin-related protein 2 / 3 complex subunit 2 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
10109 | |
Research Use Only | |
ARPC2 | |
Recombinant Protein | |
E. coli | |
MILLEVNNRI IEETLALKFE NAAAGNKPEA VEVTFADFDG VLYHISNPNG DKTKVMVSIS LKFYKELQAH GADELLKRVY GSFLVNPESG YNVSLLYDLE NLPASKDSIV HQAGMLKRNC FASVFEKYFQ FQEEGKEGEN RAVIHYRDDE TMYVESKKDR VTVVFSTVFK DDDDVVIGKV FMQEFKEGRR ASHTAPQVLF SHREPPLELK DTDAAVGDNI GYITFVLFPR HTNASARDNT INLIHTFRDY | |
Greater than 90% as determined by SDS-PAGE. |